1-(3-Brombenzyl)piperidin

Lieferant: COMBI-BLOCKS

OT-8056-1G
COBBOT-8056-1GEA 140 EUR
COBBOT-8056-1G
1-(3-Brombenzyl)piperidin
1-(3-Brombenzyl)piperidin
Formel: C₁₂H₁₆BrN
MDL: MFCD06080691
CAS-Nummer: 59507-40-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR