5-Brom-2-fluorpyrimidin

Lieferant: COMBI-BLOCKS

OT-8115-5G
COBBOT-8115-5GEA 0 EUR
COBBOT-8115-5G
5-Brom-2-fluorpyrimidin
5-Bromo-2-fluoropyrimidine
Formel: C₄H₂BrFN₂
MDL: MFCD07787364
CAS-Nummer: 62802-38-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR