4-Fluor-3-nitrophenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

PN-2049-1G
COBBPN-2049-1GEA 0 EUR
COBBPN-2049-1G
4-Fluor-3-nitrophenylboronsäurepinakolester
4-Fluor-3-nitrophenylboronsäurepinakolester
Formel: C₁₂H₁₅BFNO₄
MDL: MFCD12546515
CAS-Nummer: 1218791-09-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR