4-(2-Carboxyethyl)phenylboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
3-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)propanoic acid
PN-2055-5G
PN-2055-1G
COBBPN-2055-5GEA
1110
EUR
COBBPN-2055-5G
COBBPN-2055-1G
4-(2-Carboxyethyl)phenylboronsäurepinakolester
4-(2-Carboxyethyl)phenylboronsäurepinakolester
Formel:
C₁₅H₂₁BO₄ |
MDL:
MFCD11053827 CAS-Nummer: 797755-11-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...