4-(2-Carboxyethyl)phenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 3-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)propanoic acid

PN-2055-5G PN-2055-1G
COBBPN-2055-5GEA 1110 EUR
COBBPN-2055-5G COBBPN-2055-1G
4-(2-Carboxyethyl)phenylboronsäurepinakolester
4-(2-Carboxyethyl)phenylboronsäurepinakolester
Formel: C₁₅H₂₁BO₄
MDL: MFCD11053827
CAS-Nummer: 797755-11-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR