3,4-Dimethoxyphenylboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
3,4-Dimethoxyphenylboronic acid Pinacol ester, 1,2-Dimethoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene, 2-(3,4-Dimethoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PN-2103-5G
PN-2103-25G
PN-2103-1G
COBBPN-2103-5GEA
127
EUR
COBBPN-2103-5G
COBBPN-2103-25G
COBBPN-2103-1G
3,4-Dimethoxyphenylboronsäurepinakolester
3,4-Dimethoxyphenylboronsäurepinakolester
Formel:
C₁₄H₂₁BO₄ Molecular Weight: 264.13 g/mol |
MDL:
MFCD05863909 CAS-Nummer: 365564-10-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...