4-(Ethoxycarbonyl)phenylboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
Ethyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate, 4-(Ethoxycarbonyl)benzeneboronic acid pinacol ester, Ethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
PN-2113-1G
PN-2113-5G
COBBPN-2113-1GEA
0
EUR
COBBPN-2113-1G
COBBPN-2113-5G
4-(Ethoxycarbonyl)phenylboronsäurepinakolester
4-(Ethoxycarbonyl)phenylboronsäurepinakolester
Formel:
C₁₅H₂₁BO₄ |
MDL:
MFCD03453659 CAS-Nummer: 195062-62-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...