2-(Methoxycarbonyl)phenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-(2-Methoxycarbonylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid methyll ester, 2-(Methoxycarbonyl)phenylboronic acid Pinacol ester, Methyl 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate, 2-Methoxycarbonylphenylboronic acid pinacol ester

PN-2125-5G PN-2125-25G PN-2125-1G
COBBPN-2125-5GEA 121 EUR
COBBPN-2125-5G COBBPN-2125-25G COBBPN-2125-1G
2-(Methoxycarbonyl)phenylboronsäurepinakolester
2-(Methoxycarbonyl)phenylboronsäurepinakolester
Formel: C₁₄H₁₉BO₄
Molecular Weight: 262.1093 g/mol
MDL: MFCD05663866
CAS-Nummer: 653589-95-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR