2-(Ethoxycarbonyl)phenylboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
2-Ethoxycarbonylphenylboronic acid pinacol ester, 2-(Ethoxycarbonyl)benzeneboronic acid pinacol ester, Ethyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate, Ethyl 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
PN-2126-5G
PN-2126-1G
COBBPN-2126-5GEA
0
EUR
COBBPN-2126-5G
COBBPN-2126-1G
2-(Ethoxycarbonyl)phenylboronsäurepinakolester
2-(Ethoxycarbonyl)phenylboronsäurepinakolester
Formel:
C₁₅H₂₁BO₄ |
MDL:
MFCD03453660 CAS-Nummer: 269409-99-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...