2-Chlor-4-pyridinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-Chloropyridine-4-boronic acid pinacol ester, 2-(2-Chloro-4-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-Chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 2-Chloropyridine-4-boronic acid Pinacol ester

PN-2272-5G PN-2272-1G
COBBPN-2272-5GEA 312 EUR
COBBPN-2272-5G COBBPN-2272-1G
2-Chlor-4-pyridinboronsäurepinakolester
2-Chlor-4-pyridinboronsäurepinakolester
Formel: C₁₁H₁₅BClNO₂
Molecular Weight: 239.51 g/mol
MDL: MFCD04039870
CAS-Nummer: 458532-84-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR