3-(Trifluormethyl)phenylboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
4,4,5,5-Tetramethyl-2-[3-(trifluoromethyl)phenyl]-1,3,2-dioxaborolane, 3-(Trifluoromethyl)phenylboronic acid Pinacol ester, 4,4,5,5-Tetramethyl-2-(3-(trifluoromethyl)phenyl)-1,3,2-dioxaborolane, 1-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-3-(trifluoromethyl)benzene
PN-2378-5G
PN-2378-1G
COBBPN-2378-5GEA
166
EUR
COBBPN-2378-5G
COBBPN-2378-1G
3-(Trifluormethyl)phenylboronsäurepinakolester
3-(Trifluormethyl)phenylboronsäurepinakolester
Formel:
C₁₃H₁₆BF₃O₂ Molecular Weight: 272.0712 g/mol |
MDL:
MFCD05863925 CAS-Nummer: 325142-82-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...