4-Methoxycarbonyl-2-nitrophenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

PN-2461-5G PN-2461-25G PN-2461-1G
COBBPN-2461-5GEA 0 EUR
COBBPN-2461-5G COBBPN-2461-25G COBBPN-2461-1G
4-Methoxycarbonyl-2-nitrophenylboronsäurepinakolester
4-Methoxycarbonyl-2-nitrophenylboronsäurepinakolester
Formel: C₁₄H₁₈BNO₆
MDL: MFCD09878423
CAS-Nummer: 957065-97-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR