2-Methyl-5-nitrophenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

PN-2473-5G PN-2473-1G
COBBPN-2473-5GEA 0 EUR
COBBPN-2473-5G COBBPN-2473-1G
2-Methyl-5-nitrophenylboronsäurepinakolester
2-Methyl-5-nitrophenylboronsäurepinakolester
Formel: C₁₃H₁₈BNO₄
MDL: MFCD09027081
CAS-Nummer: 957062-84-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR