3,5-Dimethoxyphenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-(3,5-Dimethoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1,3-Dimethoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene, 3,5-Dimethoxyphenylboronic acid Pinacol ester

PN-2622-5G PN-2622-1G
COBBPN-2622-5GEA 236 EUR
COBBPN-2622-5G COBBPN-2622-1G
3,5-Dimethoxyphenylboronsäurepinakolester
3,5-Dimethoxyphenylboronsäurepinakolester
Formel: C₁₄H₂₁BO₄
Molecular Weight: 264.13 g/mol
MDL: MFCD05865191
CAS-Nummer: 365564-07-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR