2,6-Difluorphenylboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
2,6-Difluorobenzeneboronic acid pinacol ester, 2,6-Difluorophenylboronic acid, pinacol ester, 2-(2,6-Difluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PN-2659-1G
PN-2659-5G
COBBPN-2659-1GEA
0
EUR
COBBPN-2659-1G
COBBPN-2659-5G
2,6-Difluorphenylboronsäurepinakolester
2,6-Difluorphenylboronsäurepinakolester
Formel:
C₁₂H₁₅BF₂O₂ Molecular Weight: 240.06 g/mol |
MDL:
MFCD09801043 CAS-Nummer: 863868-37-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...