2,4-Dimethoxyphenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-(2,4-Dimethoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

PN-2667-5G PN-2667-1G
COBBPN-2667-5GEA 196 EUR
COBBPN-2667-5G COBBPN-2667-1G
2,4-Dimethoxyphenylboronsäurepinakolester
2,4-Dimethoxyphenylboronsäurepinakolester
Formel: C₁₄H₂₁BO₄
Molecular Weight: 264.13 g/mol
MDL: MFCD06795688
CAS-Nummer: 214360-69-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR