2-Brom-3-pyridinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

PN-2705-1G
COBBPN-2705-1GEA 0 EUR
COBBPN-2705-1G
2-Brom-3-pyridinboronsäurepinakolester
2-Brom-3-pyridinboronsäurepinakolester
Formel: C₁₁H₁₅BBrNO₂
MDL: MFCD06798246
CAS-Nummer: 452972-12-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR