2-Brom-4-pyridinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-Bromo-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 2-(2-Bromo-4-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-Bromopyridine-4-boronic acid pinacol ester, 2-Bromo-4-pyridylboronic acid Pinacol ester

PN-2708-1G
COBBPN-2708-1GEA 196 EUR
COBBPN-2708-1G
2-Brom-4-pyridinboronsäurepinakolester
2-Brom-4-pyridinboronsäurepinakolester
Formel: C₁₁H₁₅BBrNO₂
Molecular Weight: 283.96 g/mol
MDL: MFCD06798252
CAS-Nummer: 458532-82-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR