4-(Methylcarbamoyl)phenylboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
N-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
PN-3006-5G
PN-3006-1G
COBBPN-3006-5GEA
0
EUR
COBBPN-3006-5G
COBBPN-3006-1G
4-(Methylcarbamoyl)phenylboronsäurepinakolester
4-(Methylcarbamoyl)phenylboronsäurepinakolester
Formel:
C₁₄H₂₀BNO₃ |
MDL:
MFCD06795690 CAS-Nummer: 214360-57-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...