3-Chlor-2-fluor-4-pyridinboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
3-Chloro-2-fluoropyridine-4-boronic acid pinacol ester
PN-3591-250M
COBBPN-3591-250MEA
181
EUR
COBBPN-3591-250M
3-Chlor-2-fluor-4-pyridinboronsäurepinakolester
3-Chlor-2-fluor-4-pyridinboronsäurepinakolester
Formel:
C₁₁H₁₄BClFNO₂ Molecular Weight: 257,5 g/mol |
MDL:
MFCD09037476 CAS-Nummer: 1073353-71-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...