3-Chlor-2-methoxy-4-pyridinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 3-Chloro-2-methoxypyridine-4-boronic acid pinacol ester

PN-3843-1G
COBBPN-3843-1GEA 410 EUR
COBBPN-3843-1G
3-Chlor-2-methoxy-4-pyridinboronsäurepinakolester
3-Chlor-2-methoxy-4-pyridinboronsäurepinakolester
Formel: C₁₂H₁₇BClNO₃
Molecular Weight: 269,53 g/mol
MDL: MFCD06798258
CAS-Nummer: 1073353-73-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR