5-Brom-2-methoxy-3-pyridinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 5-Bromo-2-methoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 5-Bromo-2-methoxypyridine-3-boronic acid, pinacol ester

PN-3860-1G
COBBPN-3860-1GEA 149 EUR
COBBPN-3860-1G
5-Brom-2-methoxy-3-pyridinboronsäurepinakolester
5-Brom-2-methoxy-3-pyridinboronsäurepinakolester
Formel: C₁₂H₁₇BBrNO₃
Molecular Weight: 313,99 g/mol
MDL: MFCD07781157
CAS-Nummer: 1073353-75-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR