2-Methoxy-4-pyridinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-Methoxy-4-pyridylboronic acid Pinacol ester, 2-Methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 2-(2-Methoxy-4-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-Methoxypyridine-4-boronic acid pinacol ester

PN-3880-1G
COBBPN-3880-1GEA 196 EUR
COBBPN-3880-1G
2-Methoxy-4-pyridinboronsäurepinakolester
2-Methoxy-4-pyridinboronsäurepinakolester
Formel: C₁₂H₁₈BNO₃
Molecular Weight: 235,09 g/mol
MDL: MFCD07368867
CAS-Nummer: 408502-23-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR