4-(2-Hydroxy-3-(piperidin-1-yl)propoxy)phenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

PN-3889-1G
COBBPN-3889-1GEA 692 EUR
COBBPN-3889-1G
4-(2-Hydroxy-3-(piperidin-1-yl)propoxy)phenylboronsäurepinakolester
4-(2-Hydroxy-3-(piperidin-1-yl)propoxy)phenylboronsäurepinakolester
Formel: C₂₀H₃₂BNO₄
MDL: MFCD09258654
CAS-Nummer: 957061-08-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR