4-(3-(Piperidin-1-yl)propoxyphenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 1-(3-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenoxy)propyl)piperidine

PN-3910-1G
COBBPN-3910-1GEA 764 EUR
COBBPN-3910-1G
4-(3-(Piperidin-1-yl)propoxyphenylboronsäurepinakolester
4-(3-(Piperidin-1-yl)propoxyphenylboronsäurepinakolester
Formel: C₂₀H₃₂BNO₃
Molecular Weight: 345,280 g/mol
MDL: MFCD09475821
CAS-Nummer: 401895-68-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR