5-Chlor-3-pyridinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 5-Chloropyridine-3-boronic acid Pinacol ester, 2-(5-Chloro-3-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-Chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

PN-5001-1G
COBBPN-5001-1GEA 210 EUR
COBBPN-5001-1G
5-Chlor-3-pyridinboronsäurepinakolester
5-Chlor-3-pyridinboronsäurepinakolester
Formel: C₁₁H₁₅BClNO₂
Molecular Weight: 239.51 g/mol
MDL: MFCD06798284
CAS-Nummer: 865186-94-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR