7-Brom-2-indolboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 7-Bromo-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole

PN-5039-1G
COBBPN-5039-1GEA 595 EUR
COBBPN-5039-1G
7-Brom-2-indolboronsäurepinakolester
7-Brom-2-indolboronsäurepinakolester
Formel: C₁₄H₁₇BBrNO₂
Molecular Weight: 322,0053 g/mol
MDL: MFCD11858379
CAS-Nummer: 1072812-23-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR