4-Methoxy-2-nitrophenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

PN-5083-1G PN-5083-250M
COBBPN-5083-1GEA 0 EUR
COBBPN-5083-1G COBBPN-5083-250M
4-Methoxy-2-nitrophenylboronsäurepinakolester
4-Methoxy-2-nitrophenylboronsäurepinakolester
Formel: C₁₃H₁₈BNO₅
MDL: MFCD10699701
CAS-Nummer: 1073353-81-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR