2-Acetamido-5-chlorphenylboronsäurepinakolester
Lieferant: COMBI-BLOCKS
PN-5084-1G
COBBPN-5084-1GEA
0
EUR
COBBPN-5084-1G
2-Acetamido-5-chlorphenylboronsäurepinakolester
2-Acetamido-5-chlorphenylboronsäurepinakolester
Formel:
C₁₄H₁₉BClNO₃ |
MDL:
MFCD09878425 CAS-Nummer: 957063-08-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...