5-Methoxy-3-pyridinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 5-Methoxy-3-pyridylboronic acid Pinacol ester, 3-Methoxypyridine-5-boronic acid Pinacol ester, 3-Methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 2-(5-Methoxy-3-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

PN-5109-5G PN-5109-1G
COBBPN-5109-5GEA 181 EUR
COBBPN-5109-5G COBBPN-5109-1G
5-Methoxy-3-pyridinboronsäurepinakolester
5-Methoxy-3-pyridinboronsäurepinakolester
Formel: C₁₂H₁₈BNO₃
Molecular Weight: 235.0872 g/mol
MDL: MFCD03412068
CAS-Nummer: 445264-60-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR