2-Hydroxy-5-pyridinboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-ol, 6-Hydroxypyridine-3-boronic acid pinacol ester
PN-5130-1G
PN-5130-5G
COBBPN-5130-1GEA
127
EUR
COBBPN-5130-1G
COBBPN-5130-5G
2-Hydroxy-5-pyridinboronsäurepinakolester
2-Hydroxy-5-pyridinboronsäurepinakolester
Formel:
C₁₁H₁₆BNO₃ Molecular Weight: 221.06 g/mol |
MDL:
MFCD06795671 CAS-Nummer: 1054483-78-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...