2-Fluor-4-nitrophenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

PN-5185-1G PN-5185-250M
COBBPN-5185-1GEA 0 EUR
COBBPN-5185-1G COBBPN-5185-250M
2-Fluor-4-nitrophenylboronsäurepinakolester
2-Fluor-4-nitrophenylboronsäurepinakolester
Formel: C₁₂H₁₅BFNO₄
MDL: MFCD09264075
CAS-Nummer: 1073353-89-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR