3-(Piperidin-1-ylmethyl)phenylboronsäurepinakolester Hydrochlorid

Lieferant: COMBI-BLOCKS

Synonyme: 1-{[3-(Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl}piperidin-1-ium chloride

PN-5194-1G
COBBPN-5194-1GEA 149 EUR
COBBPN-5194-1G
3-(Piperidin-1-ylmethyl)phenylboronsäurepinakolester Hydrochlorid
3-(Piperidin-1-ylmethyl)phenylboronsäurepinakolester Hydrochlorid
Formel: C₁₈H₂₉BClNO₂
MDL: MFCD06657902
CAS-Nummer: 1021186-08-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR