4-(1-Boc-piperidin-4-yloxy)-3-methoxyphenylboronsäurepinakolester
Lieferant: COMBI-BLOCKS
PN-5215-100M
COBBPN-5215-100MEA
742
EUR
COBBPN-5215-100M
4-(1-Boc-piperidin-4-yloxy)-3-methoxyphenylboronsäurepinakolester
4-(1-Boc-piperidin-4-yloxy)-3-methoxyphenylboronsäurepinakolester
Formel:
C₂₃H₃₆BNO₆ |
MDL:
MFCD16618965 CAS-Nummer: 1246372-53-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...