5-Chlor-2-methyl-4-(trifluormethyl)phenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

PN-5223-100M
COBBPN-5223-100MEA 864 EUR
COBBPN-5223-100M
5-Chlor-2-methyl-4-(trifluormethyl)phenylboronsäurepinakolester
5-Chlor-2-methyl-4-(trifluormethyl)phenylboronsäurepinakolester
Formel: C₁₄H₁₇BClF₃O₂
MDL: MFCD11053829
CAS-Nummer: 1073353-97-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR