2,5-Dichlor-4-pyridinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2,5-Dichloropyridine-4-boronic acid pinacol ester, 2,5-Dichloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

PN-5232-1G
COBBPN-5232-1GEA 298 EUR
COBBPN-5232-1G
2,5-Dichlor-4-pyridinboronsäurepinakolester
2,5-Dichlor-4-pyridinboronsäurepinakolester
Formel: C₁₁H₁₄BCl₂NO₂
Molecular Weight: 273,95 g/mol
MDL: MFCD06798256
CAS-Nummer: 1073353-98-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR