3-Cyan-2-methoxy-5-pyridinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-Methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)nicotinonitrile

PN-5386-1G
COBBPN-5386-1GEA 613 EUR
COBBPN-5386-1G
3-Cyan-2-methoxy-5-pyridinboronsäurepinakolester
3-Cyan-2-methoxy-5-pyridinboronsäurepinakolester
Formel: C₁₃H₁₇BN₂O₃
Molecular Weight: 260,100 g/mol
MDL: MFCD11504968
CAS-Nummer: 1073354-05-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR