2,3-Dimethoxy-5-pyridinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2,3-Dimethoxypyridine-5-boronic acid, pinacol ester, 2,3-Dimethoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

PN-5388-1G
COBBPN-5388-1GEA 595 EUR
COBBPN-5388-1G
2,3-Dimethoxy-5-pyridinboronsäurepinakolester
2,3-Dimethoxy-5-pyridinboronsäurepinakolester
Formel: C₁₃H₂₀BNO₄
Molecular Weight: 265,12 g/mol
MDL: MFCD11857651
CAS-Nummer: 1083168-92-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR