2,6-Dichlor-4-pyridinboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
2,6-Dichloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 2,6-Dichloropyridine-4-boronic acid Pinacol ester, 2,6-Dichloropyridine-4-boronic acid, pinacol ester, 2-(2,6-Dichloro-4-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PN-5391-1G
COBBPN-5391-1GEA
88.6
EUR
COBBPN-5391-1G
2,6-Dichlor-4-pyridinboronsäurepinakolester
2,6-Dichlor-4-pyridinboronsäurepinakolester
Formel:
C₁₁H₁₄BCl₂NO₂ Molecular Weight: 273,95 g/mol |
MDL:
MFCD09746195 CAS-Nummer: 408492-27-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...