2-Methoxy-3-(methoxycarbonyl)-5-pyridinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 2-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)nicotinate, 2-Methoxy-3-(carbomethoxy)pyridine-5-boronic acid, pinacol ester

PN-5463-1G
COBBPN-5463-1GEA 761 EUR
COBBPN-5463-1G
2-Methoxy-3-(methoxycarbonyl)-5-pyridinboronsäurepinakolester
2-Methoxy-3-(methoxycarbonyl)-5-pyridinboronsäurepinakolester
Formel: C₁₄H₂₀BNO₅
MDL: MFCD11855973
CAS-Nummer: 1083168-93-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR