3-Methoxy-5-(methoxycarbonyl)phenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 3-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate, 3-Methoxy-5-(methoxycarbonyl)phenylboronic acid, pinacol ester

PN-5505-1G PN-5505-250M
COBBPN-5505-1GEA 0 EUR
COBBPN-5505-1G COBBPN-5505-250M
3-Methoxy-5-(methoxycarbonyl)phenylboronsäurepinakolester
3-Methoxy-5-(methoxycarbonyl)phenylboronsäurepinakolester
Formel: C₁₅H₂₁BO₅
Molecular Weight: 292.135 g/mol
MDL: MFCD12026074
CAS-Nummer: 889654-06-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR