5-Acetyl-2,3-(methylenedioxy)phenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

PN-5512-1G
COBBPN-5512-1GEA 0 EUR
COBBPN-5512-1G
5-Acetyl-2,3-(methylenedioxy)phenylboronsäurepinakolester
5-Acetyl-2,3-(methylenedioxy)phenylboronsäurepinakolester
Formel: C₁₅H₁₉BO₅
MDL: MFCD11855978
CAS-Nummer: 1150271-59-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR