2-(Ethoxycarbonyl)furan-5-boronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: Ethyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)furan-2-carboxylate

PN-5514-1G
COBBPN-5514-1GEA 635 EUR
COBBPN-5514-1G
2-(Ethoxycarbonyl)furan-5-boronsäurepinakolester
2-(Ethoxycarbonyl)furan-5-boronsäurepinakolester
Formel: C₁₃H₁₉BO₅
Molecular Weight: 266,10 g/mol
MDL: MFCD11855980
CAS-Nummer: 1073338-92-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR