5-(tert-Butyldimethylsilyloxy)-2,3-difluorphenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

PN-5535-1G
COBBPN-5535-1GEA 303 EUR
COBBPN-5535-1G
5-(tert-Butyldimethylsilyloxy)-2,3-difluorphenylboronsäurepinakolester
5-(tert-Butyldimethylsilyloxy)-2,3-difluorphenylboronsäurepinakolester
Formel: C₁₈H₂₉BF₂O₃Si
MDL: MFCD11855981
CAS-Nummer: 1150271-62-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR