4-Chlor-3,5-dimethylphenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 3,5-Dimethyl-4-chlorophenylboronic acid pinacol ester, 2-(4-Chloro-3,5-dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

PN-5541-5G PN-5541-1G PN-5541-250M
COBBPN-5541-5GEA 1490 EUR
COBBPN-5541-5G COBBPN-5541-1G COBBPN-5541-250M
4-Chlor-3,5-dimethylphenylboronsäurepinakolester
4-Chlor-3,5-dimethylphenylboronsäurepinakolester
Formel: C₁₄H₂₀BClO₂
Molecular Weight: 266.571 g/mol
MDL: MFCD12405360
CAS-Nummer: 1111096-20-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR