2-Amino-3-(trifluormethyl)-5-pyridinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-3-(trifluoromethyl)pyridin-2-amine

PN-5555-1G
COBBPN-5555-1GEA 0 EUR
COBBPN-5555-1G
2-Amino-3-(trifluormethyl)-5-pyridinboronsäurepinakolester
2-Amino-3-(trifluormethyl)-5-pyridinboronsäurepinakolester
Formel: C₁₂H₁₆BF₃N₂O₂
MDL: MFCD12923414
CAS-Nummer: 947249-01-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR