4-Chlor-2-fluor-5-methylphenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 4-Chloro-2-fluoro-5-methylphenylboronic acid, pinacol ester

PN-5634-1G
COBBPN-5634-1GEA 1240 EUR
COBBPN-5634-1G
4-Chlor-2-fluor-5-methylphenylboronsäurepinakolester
4-Chlor-2-fluor-5-methylphenylboronsäurepinakolester
Formel: C₁₃H₁₇BClFO₂
MDL: MFCD11855985
CAS-Nummer: 1126320-27-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR