4-Chlor-2-fluor-5-(methoxycarbonyl)phenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

PN-5667-1G
COBBPN-5667-1GEA 0 EUR
COBBPN-5667-1G
4-Chlor-2-fluor-5-(methoxycarbonyl)phenylboronsäurepinakolester
4-Chlor-2-fluor-5-(methoxycarbonyl)phenylboronsäurepinakolester
Formel: C₁₄H₁₇BClFO₄
MDL: MFCD12026085
CAS-Nummer: 1073339-13-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR