2,6-Dimethoxy-4-pyridinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

PN-5680-1G
COBBPN-5680-1GEA 1430 EUR
COBBPN-5680-1G
2,6-Dimethoxy-4-pyridinboronsäurepinakolester
2,6-Dimethoxy-4-pyridinboronsäurepinakolester
Formel: C₁₃H₂₀BNO₄
MDL: MFCD11855988
CAS-Nummer: 1150561-54-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR