5-Cyan-2-fluor-3-methoxyphenylboronsäurepinakolester
Lieferant: COMBI-BLOCKS
PN-5683-1G
COBBPN-5683-1GEA
1630
EUR
COBBPN-5683-1G
5-Cyan-2-fluor-3-methoxyphenylboronsäurepinakolester
5-Cyan-2-fluor-3-methoxyphenylboronsäurepinakolester
Formel:
C₁₄H₁₇BFNO₃ |
MDL:
MFCD12026087 CAS-Nummer: 1150561-55-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...